Polyester labels for M611 and BMP™61 printers, type B-481, StainerBondz™
Supplier: Brady
BRDY150865EA 64 EUR
BRDY150865 BRDY150864
Polyester labels for M611 and BMP™61 printers, type B-481, StainerBondz™
Die-cut labels are made from StainerBondz™ polyester, with a permanent acrylic adhesive and matte finish.
  • Die-cut labels are supplied on a roll
  • Label thickness is 0,084 mm
  • Maximum service temperature: 130 °C
  • Minimum service temperature: –80 °C
  • Ribbon series: R6400
Super chemical-resistant polyester (B-481) labels are designed for use in the slide staining process and other laboratory identifications requiring superior chemical resistance. Performs well in the commonly used H&E (hematoxilin and eosin) slide staining process.