Isohexane (mixture of isomers)
Supplier: Roth Carl
ROTHT904.1EA 126 EUR
ROTHT904.1
Isohexane (mixture of isomers)
Isohexane (mixture of isomers)
Formula: C₆H₁₄
MW: 86,18 g/mol
Boiling Pt: 60 °C (1013 hPa)
Melting Pt: –154 °C
Density: 0,653 g/cm³ (25 °C)
Flash Pt: –33 °C
MDL Number: MFCD00009406
UN: 1208
ADR: 3,II