You Searched For: Tube Furnaces
5  results were found
SearchResultCount:"5"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: NABERTHERM GMBH
Description: These compact bench top tube furnaces with integrated controllers can be widely used for a variety of processes. Equipped with a standard C 530 cerami...
Supplier: NABERTHERM GMBH
Description: These compact tube furnaces are used when laboratory experiments must be performed horizontally, vertically, or at specific angles. The ability to con...
Supplier: NABERTHERM GMBH
Description: These compact tube furnaces with a SiC rod heating and integrated switchgear with controller can be used universally for many processes. They represen...
Supplier: CARBOLITE
Description: The vertical split tube furnace range VST (single zone) and TVS (three zone) uses free radiating wire elements embedded within the insulation of the f...
Catalog Number: (CARBPTC1220-230SN)
Supplier: CARBOLITE
Description: Tube Furnace, PTC 12/20/150, Max. temp.: 1200 °C, Heated length: 150 mm, Ø int. tube: 20 mm, Power: Max. 1100 W, W×D×H ext.: 311×229×400 mm
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 5 of 5
no targeter for Bottom