You Searched For: Inoculating Loops and Needles
39  results were found
SearchResultCount:"39"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Inoculating loops and needles are devices used for inoculation, serial dilution, sterile sampling, transfer, and microbiological sample spreading.
New Product
Supplier: VWR Collection
Description: PS, sterile. These loops ensure consistent wetting and complete liquid transfer. Available rigid or flexible and have smooth edges to avoid gouging th...
Catalog Number: (612-9360)
Supplier: VWR Collection
Description: PS, sterile.
UOM: 1 * 1.000 items
Supplier: VWR Collection
Description: PS, sterile. Disposable plastic inoculating needles.
Catalog Number: (612-2496)
Supplier: VWR Collection
Description: PS, sterile. Calibrated, disposable inoculating loops made from flexible or rigid plastic that cater for microbiologist's different applications and m...
UOM: 1 * 1.000 items
Supplier: COPAN
Description: Inoculating loop, 1 µl, Sterile
Supplier: COPAN
Description: These nickel-chrome metal wire loops are available non calibrated in a range of sizes with internal diameters (Ø) of 2, 3, 4 or 5 mm, or calibrated, 1...
Supplier: RSG
Description: These NiCr 8020 loops are available in a range of sizes and lengths with internal diameter of 1,5, 2,5 or 4 mm. They are heat resistant and can be fla...
Supplier: 3M Food Safety
Description: This 3Mâ„¢ Petrifilmâ„¢ Spreader is an easy-to-use tool designed to quickly and evenly distribute inoculum within the 3Mâ„¢ Petrifilmâ„¢ Plate.
Supplier: COPAN
Description: A range of nickel-chrome metal wire loops attached to lightweight insulated aluminium handles. Combo-loops are non calibrated and available in a range...
Supplier: ANICRIN S.R.L.
Description: Inoculating loop, Blue, 10 µl
Supplier: BOCHEM
Description: Stainless steel 18/10. Length 100 mm.
Supplier: Simport Scientific
Description: PS, sterile. Loops and needles are smooth and flexible to facilitate uniform streaking without damaging the gel surface.
Supplier: KARTELL
Description: Inoculating loop, L-shaped, PP, Sterile
Supplier: Corning
Description: These polystyrene inoculating loops are highly ergonomic and are color coded for easy identification.
Supplier: Brand
Description: PS, gamma sterilised, disposable. For inoculation of nutrient media. Available either with loop at one end and needle at the other or loops at both en...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 39
no targeter for Bottom