You Searched For: Desiccators, Plastic
15  results were found
SearchResultCount:"15"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Thermo Fisher Scientific
Description: Transparent PC lid and blue PP base with or without PTFE stopcock. Accepts 6,4 mm (¹/₄") int.Ø. vacuum tubing and can be used with any standard 230 mm...
Catalog Number: (467-3120)
Supplier: Thermo Fisher Scientific
Description: Transparent PC, unbreakable and lightweight. Suitable for vacuum and non vacuum applications. Has wide base for greater stability. Accepts 6,4 mm (¹/₄...
UOM: 1 * 1 items
Catalog Number: (467-0021)
Supplier: Thermo Fisher Scientific
Description: Ideal for drying and drying processes. Cover is transparent, robust PC with recessed rim. Base is durable blue PP.
UOM: 1 * 1 items
Supplier: Bel-Art Products, a Part of SP
Description: Desiccators with white PP base and clear PC domed lid with large ribbed knob. Desiccant can be placed on or under the PP plate.
Supplier: KARTELL
Description: Transparent polycarbonate cover with polypropylene base. The specially grooved stopper ensures air is readmitted very slowly to avoid disturbing the c...
Catalog Number: (BELAF420290001)
Supplier: Bel-Art Products, a Part of SP
Description: Clear polycarbonate desiccator with large storage space, portable and easy to use. Two gas ports accept tubing (inner Ø 6,35 mm) and have easy turning...
UOM: 1 * 1 items
Supplier: Bel-Art Products, a Part of SP
Description: These compact, high-quality vacuum desiccators in clear or amber (UV blocking) polycarbonate maintain a vacuum and stay gas-tight allowing reliable dr...
Supplier: Bel-Art Products, a Part of SP
Description: Degas liquids quickly and easily in the laboratory or classroom with this small plastic vacuum desiccator. These strong, shatterproof units provide dr...
Catalog Number: (BELAF420290000)
Supplier: Bel-Art Products, a Part of SP
Description: This domed desiccator provides a spacious interior for large, moisture sensitive materials. Laboratory grade polycarbonate provides a clear, unobstruc...
UOM: 1 * 1 items
Catalog Number: (VITL326294)
Supplier: VITLAB
Description: Ideal dessicators for use in educational laboratories.
UOM: 1 * 1 items
Supplier: Brand
Description: Dessicators with polycarbonate lid, PP base and desiccant tray. Venting stopper with non-return valve in lid. Seal between lid and base: polychlorobut...
Catalog Number: (467-2245)
Supplier: Bel-Art Products, a Part of SP
Description: Desiccators with top made of clear polycarbonate, for easy viewing of contents, and white polypropylene bottom. The small size of this desiccator allo...
UOM: 1 * 1 items
Supplier: SciLabware
Description: Transparent polycarbonate lid and base.
Catalog Number: (BELAF420430000)
Supplier: Bel-Art Products, a Part of SP
Description: Ideal for vacuum drying, degassing and teaching demonstrations, this vacuum chamber can withstand a full vacuum at room temperature.
UOM: 1 * 1 items
Supplier: Thermo Fisher Scientific
Description: Vacuum chamber systems include a transparent jar and neoprene gasketed vacuum plate with a tubing adapter that fits 6,4 mm Int.Ø tubing. Jars are avai...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 15 of 15
no targeter for Bottom