You Searched For: Magnetic Stirrers, Multi-position
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: 2MAG
Description: The hermetically sealed stainless steel housing is water, germ and dust-proof. Operating temperatures for basic version up to 50 °C, for high temperat...
Supplier: VELP SCIENTIFIC
Description: Illuminated magnetic stirrers with single-position (AMI) or four separately controlled positions (AMI 4). Specially designed for titrations, and parti...
Supplier: VELP SCIENTIFIC
Description: Magnetic stirrer able to stir up to six beakers having a maximum diameter of 85 mm (MULTISTIRRER 6) or up to 15 beakers having a maximum diameter of 6...
Supplier: 2MAG
Description: Stirring systems for careful mixing of cell cultures. Suitable for culture flasks with spinning propeller and spinning ball systems. The hermetically ...
Supplier: 2MAG
Description: Multi-position magnetic stirrers with hermetically sealed stainless steel housing which is water-, germ- and dust-proof. Suitable for shaking micotitr...
Supplier: 2MAG
Description: Ultra-flat magnetic stirrers with hermetically sealed stainless steel housing. Operating temperatures up to 50 °C.
Supplier: VELP SCIENTIFIC
Description: The MULTISTIRRER Digital are multi-position magnetic stirrers with a resistant stainless-steel top plate, that remains cold even after several days of...
Supplier: IKA
Description: Compact digital magnetic stirrers with 5, 10 or 15 stirring positions, designed for synchronous stirring and suitable for continuous operation. The we...
Supplier: H + P LABORTECHNIK
Description: Compact magnetic stirrers with VARIOMAG drive, hermetically sealed in resistant stainless steel housing (except MINI units which are plastic), making ...
Catalog Number: (SCIISI-3002)
Supplier: SCIENTIFIC INDUSTRIES (ELMIS)
Description: The programmable revolutionary 4-position MultiMagStir Genie provides low to high speed stirring and editable memory profiles.
UOM: 1 * 1 items
Supplier: Cole Parmer
Description: Three independent stirring or heating/stirring positions yet only measures L×W: 600×270 mm.
Supplier: H + P LABORTECHNIK
Description: Single and multi-position stirrers for up to four vessels. Biosystem models and accessory Biomodul 40 B controller are suitable for mixing culture bro...
Catalog Number: (442-3133)
Supplier: H + P LABORTECHNIK
Description: Multi-position stirrers with direct control for up to 15 vessels, are ideal for general laboratory and mass screening applications. Units provide smoo...
UOM: 1 * 1 items
Catalog Number: (SCIISI-5256)
Supplier: SCIENTIFIC INDUSTRIES (ELMIS)
Description: Stirrer, Multi-position Magnetic Stirrer, Genie®, EU-plug, Quadmag Genie®, digital, Max. stirring capacity per stirrer H₂O: 2000 ml, Speed range: 250 - 1300 min⁻¹
UOM: 1 * 1 items
Supplier: H + P LABORTECHNIK
Description: Multi-position magnetic stirrers for reliable, safe performance with pre-selectable power settings, which optimise stirring efficiency for different l...
Supplier: BARNSTEAD (INC THERMOLYNE)
Description: Carry out solubility studies, digestion, titration and enzymatic studies with Thermo Scientific™ Super-Nuova™ Multi-Position Digital Stirrers. Offerin...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom