You Searched For: Magnetic Hotplate Stirrers, Multi-position
6  results were found
SearchResultCount:"6"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VELP SCIENTIFIC
Description: Safe and compact digital multi-position magnetic hotplate stirrers available with 6 or 15 positions. The MULTI-HS include features such as timer, safe...
Supplier: 2MAG
Description: Hotplate stirrers that have 6 or 15 powerful stirring positions. The units have a clear digital display for settings of stirrer speed, stirrer power a...
Supplier: VELP SCIENTIFIC
Description: Digital magnetic hotplate stirrer with four independent positions and CerAlTopâ„¢ plate, perfect for thermoregulation with PT100 and VTF connection.
Catalog Number: (442-1113)
Supplier: VELP SCIENTIFIC
Description: Multiple-position hot plate stirrer with four separately controlled stirring plates coated with aluminium alloy which provides uniform heat distributi...
UOM: 1 * 1 items
Supplier: IKA
Description: Multi-position digital magnetic hotplate stirrers with 5, 10 or 15 stirring positions, designed for synchronous heating and stirring, suitable for con...
Catalog Number: (WITGDH.WMH03506)
Supplier: WITEG LABORTECHNIK GMBH
Description: Digital multi-position magnetic stirrer ideal for mixing and heating simultaneously. Ideal for heat sensitive procedures in microbiology, biochemistry...
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 6 of 6
no targeter for Bottom