You Searched For: Standard Ovens
21  results were found
SearchResultCount:"21"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: VENTI-Line® drying ovens are designed for all standard drying and tempering tasks as well as sterilising glassware. They have a temperature range betw...
Supplier: VWR Collection
Description: DRY-Line® drying ovens are designed for all standard drying and tempering tasks as well as sterilising glassware. They have a temperature range betwee...
Sale
Supplier: MEMMERT
Description: The range includes forced convection models with a temperature range up to 300 Â°C. There is a choice of two different displays, SingleDISPLAY or ...
Supplier: MEMMERT
Description: The range includes natural convection models with a temperature range up to +300 Â°C. There is a choice of two different displays, SingleDISPLAY o...
Supplier: MEMMERT
Description: UNpa paraffin ovens provide precise and homogenous temperature control of paraffin embedding medium without any loss in quality. UNpa paraffin ovens a...
Supplier: MEMMERT
Description: The UF TS pass through ovens help to minimise contamination when transporting samples or components. They have an easy-to-clean interior, fast recover...
Supplier: Binder
Description: The Binder FP series of material test chambers with mechanical convection and program functions provide reliably short drying times and particularly f...
Catalog Number: (466-0023)
Supplier: Binder
Description: Multi-functional drying ovens with stainless steel chambers which are ideal for use in the pharmaceutical and chemical industries, biotechnology, medi...
UOM: 1 * 1 items
Supplier: Binder
Description: With natural convection, Avantgarde.Line heating and drying chambers offer quality and process reliability. With a wide temperature range up to 300 °C...
Supplier: Binder
Description: With forced convection, the Avantgarde.Line heating and drying chambers offer quality and process reliability. The wide temperature range of up to 300...
Catalog Number: (466-0488)
Supplier: Binder
Description: These drying ovens for limited quantities of solvents, have high temperature accuracy and silicone- and dust-free stainless steel chambers. Units are ...
UOM: 1 * 1 items
Catalog Number: (466-3249)
Supplier: Binder
Description: Universal drying ovens with stainless steel chambers which are ideal for use in the pharmaceutical and chemical industries, biotechnology, medicine an...
UOM: 1 * 1 items
Supplier: Binder
Description: With forced convection and enhanced timer functions, the Avantgarde.Line heating and drying chambers offer quality and process reliability. With a wid...
Supplier: Binder
Description: Universal drying ovens with stainless steel chambers which are ideal for drying and sterilisation applications in the pharmaceutical and chemical indu...
Catalog Number: (466-3161)
Supplier: Binder
Description: Robust space-saving oven/steriliser with hydraulic mechanical thermostat and temperature setting via analogue dial indicator thermometer.
UOM: 1 * 1 items
Supplier: THERMO BRAND
Description: Herathermâ„¢ large capacity ovens were designed for larger samples or high sample volume. All models have a large, easy to view, vacuum fluorescence dis...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 21
no targeter for Bottom