You Searched For: Testing Chambers
19  results were found
SearchResultCount:"19"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: The VWR® Constant climate chambers with Peltier technology feature excellent variation and fluctuation values for temperature and humidity. They are e...
Energy Efficient
Supplier: Binder
Description: The KBF P constant climatic chambers with ICH-compliant UV/Vis light source and KBF LQC with light dose control have a unique light concept that guara...
Supplier: Binder
Description: The WIC series walk-in-chambers offer a wide range of features and benefits to perfectly meet requirements in stability testing. Perfect test conditio...
New Product
Supplier: Binder
Description: The MKF series of dynamic climate chambers has been specially developed as precision heating and cooling chambers for industrial material testing and ...
Supplier: Binder
Description: The KBF series is ideal for reliable stability tests and precise maintenance of constant climatic conditions. Constant temperature and humidity values...
Supplier: MEMMERT
Description: Humidity chambers which have corrosion resistant, stainless steel inner chamber and housing for long-term climatic tests at constant temperature and h...
Supplier: MEMMERT
Description: The ICH range of climatic chambers offers rapid and precise temperature control from 10 up to 60 °C with humidification from 10 to 80% RH. These compr...
Supplier: Binder
Description: The MK series chambers are suitable for all heat and cold tests from –40 to 180 °C. This is an alternative to complex individual solutions for cyclica...
Supplier: Binder
Description: The KBW series plant growth chambers are precise units for plant cultivation under advanced working conditions. They provide perfect simulation of a w...
Supplier: Binder
Description: KMF constant climate chambers ensure constant test conditions throughout the stainless steel chamber even with a full load and only require a small am...
Supplier: MEMMERT
Description: Maximum energy effiency in continous operation - these powerful constant climate chambers work with the new Advanced Peltier Technology. These devices...
Energy Efficient
Supplier: Binder
Description: The KBWF series plant growth chambers are precise units for plant cultivation under the most up-to-date working conditions with optimum climatic condi...
Supplier: Binder
Description: Solid Line KBF-S climate chambers are ideal for long-term testing and accelerated stability testing of pharmaceutical products in accordance with ICH ...
Supplier: Binder
Description: The KBF-S ECO climatic chambers are very energy-efficient thanks to thermoelectric cooling technology with heat dissipation. The thermoelectric coolin...
Supplier: MEMMERT
Description: Climatic chamber, Capacity: 256 L, Forced convection, Temperature fluctuation (time): ±0,2…0,5 °C, Temperature variation (spatial): ±0,5...2 °C, Weight: 297 kg
Supplier: Binder
Description: Standard climate chambers that work with APT.lineâ„¢ preheating chamber technology.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 19
no targeter for Bottom