You Searched For: Syringes, Medical
58  results were found
SearchResultCount:"58"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: HENKE SASS WOLF GMBH
Description: Disposable, three-piece syringes with PP barrel and piston, polyisoprene stopper, latex-free.
Supplier: HENKE SASS WOLF GMBH
Description: Disposable, two-piece, without needle, PP barrel and PE piston, latex- and silicone oil-free. Syringes are also available with especially designed ora...
Supplier: BÃœRKLE
Description: Syringe, medical, SteriPlast®, PP, Sterile, Syringe with Luer adapter, transparent, sterile, Luer tip, Capacity: 100 ml, Graduation: 2 ml, Pk: 10
Catalog Number: (HSWA2450030100)
Supplier: HENKE SASS WOLF GMBH
Description: Non sterile, reusable, glass barrel and stainless steel piston. These syringes can be used for human and veterinary medicine.
UOM: 1 * 1 items
Supplier: B. Braun
Description: PP syringe with safe plunger backstop and minimal residual volume.
Catalog Number: (613-2793)
Supplier: HENKE SASS WOLF GMBH
Description: These syringes are commonly used for embryo transfer and oocyte handling.
UOM: 1 * 100 items
Catalog Number: (BDAM364356)
Supplier: BD Medical
Description: Critical care syringes can be used to measure arterial blood gas, ionised calcium, ionised magnesium, as well as perform critical care tests.
UOM: 1 * 100 items
Supplier: Poulten & Graf
Description: Barrel made from Durobax® glass, metal tip and interchangeable piston, no needle, autoclavable.
Supplier: TERUMO
Description: Sterile, disposable, three-piece, latex-free.
Supplier: SOCOREX ISBA
Description: Dosysâ„¢ 174 premium: Self-refilling, automatic syringe with detachable, Luer lock valve system. Enhanced corrosion resistance.
Supplier: B. Braun
Description: These fine dosage syringes are made of PP/PS. Plunger stopper with double sealing ring for slow aspiration and injection made of natural latex free sy...
Supplier: B. Braun
Description: PP, three-piece syringe for injection of U-100 insulin (50 or 100 IU/ml) for veterinary use. Smooth action, natural latex-free plunger stoppers with d...
Supplier: B. Braun
Description: Sterile, disposable, three-piece, made from PP, latex- and PVC-free.
Catalog Number: (720-2526)
Supplier: B. Braun
Description: PP, three-piece, single use, natural latex-free plunger stoppers with double sealing ring and highly transparent barrel.
UOM: 1 * 1 items
Supplier: B. Braun
Description: Single use syringes for manual use, with green plunger. Easy aspiration up to the maximum volume. Barrel made from PP and plunger from PE.
Supplier: EMBECTA
Description: Syringe, medical, BD Micro-Fine™, Insulin syringe, with needle, 29G, Length: 12,7 mm, Ø ext.: 0,33 mm, Capacity: 1 ml, Pk: 500
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 58
no targeter for Bottom