You Searched For: PE/LDPE/HDPE Stoppers
44  results were found
SearchResultCount:"44"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Stopper for test tubes Ø 12 to 16 mm, polyethylene, with tongues.
Catalog Number: (462-0585)
Supplier: VWR Collection
Description: Drain stoppers, VWR®
UOM: 1 * 5 items
Supplier: VWR Collection
Description: Greaseless closure, which prevents ground joints from seizing. With inclined, circumferential sealing lips for better sealing and reduced risk of brea...
Supplier: Simport Scientific
Description: Ideal closures for glass and plastic reagent, sample and culture tubes, LDPE.
Supplier: Bohlender
Description: For closing ground joint parts. With ground joint and sealing rings on the outside; knurled or hexagonal grip. Compared with glass stoppers, they are ...
Supplier: Simport Scientific
Description: HDPE. Fit tubes of 15 ml capacity and 17 mm Ø.
Supplier: Simport Scientific
Description: Designed for easy-on, easy-off use. Cap removal is simple and quick. FitsAllâ„¢ closure is ideal for recapping most 12 and 16 mm O.D. evacuated glass bl...
Supplier: Simport Scientific
Description: PE Plug type push-in caps and 2-position snap caps are made of polyethylene for test tubes with outside diameters of 12 or 17 mm. Dual position caps o...
Supplier: B. Braun
Description: These PE Combi-Stoppers are used for sealing male or female Luer-Lock and Luer-Slip connections.
Catalog Number: (GRANUWB-DS)
Supplier: GRANT INSTRUMENTS LTD.
Description: Drain stoppers, recommended when using heat transfer beads, to prevent beads entering the drain hole.
UOM: 1 * 5 items
Catalog Number: (720-2576)
Supplier: B. Braun
Description: These stoppers ensure a sterile closure of female connections after use.
UOM: 1 * 100 items
Supplier: Brand
Description: PP or PE-LD, hollow, with sealing ribs and square or octagonal grip.
Supplier: COWIE
Description: Stopper with sealing rings and self releasing ring , PTFE, NS: 24/40, Height: 65 mm
Supplier: COWIE
Description: PTFE. Stopper with flat head fit standard ground glass joints.
Supplier: KARTELL
Description: Finned plugs for disposable tubes 11 to 13 mm or 15 to 17 mm in diameter. The finned plug has been carefully designed. Available colours: Neutral (00)...
Supplier: SciLabware
Description: These PE stoppers are ideally suited for use with glass or plastic volumetric flasks and measuring cylinders etc. which have longer 'B' length sockets...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 44
no targeter for Bottom