You Searched For: Swabs in Tubes
9  results were found
SearchResultCount:"9"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: COPAN
Description: Swabs in tubes manufactured from high quality PP.
Supplier: COPAN
Description: Gamma sterilised swabs available in various combinations.
Catalog Number: (KIMA23117)
Supplier: KIMA SAS DI CHIARIN RENZO & C
Description: Swab in tube with label and plastic stick, PP, Length: 140 mm, Ø: 12 mm
UOM: 1 * 100 items
Supplier: DELTALAB
Description: Sterile dry swabs in impact resistant PP tubes.
Supplier: Simport Scientific
Description: Sterile specimen collection swab.
Catalog Number: (MEDWMW1028D)
Supplier: MEDICAL WIRE & EQUIPMENT
Description: Individual labelled sterile tubes with polyester bud, standard plastic shaft.
UOM: 1 * 800 items
Supplier: Greiner Bio-One
Description: These swab tubes are suitable for taking non-human bacteriological, serological or cytological samples.
Supplier: HERENZ, HEINZ GMBH
Description: Sterile dry swabs in tubes, without transport medium.
Supplier: LP ITALIANA SPA
Description: Sterile dry swabs in tubes.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 9 of 9
no targeter for Bottom