You Searched For: Disposable Earplugs
28  results were found
SearchResultCount:"28"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: 3M
Description: Soft, smooth, dirt resistant foam earplugs mould to the unique shape of each ear for excellent hearing protection.
Catalog Number: (MMMA393-2026-50)
Supplier: 3M
Description: E-A-Râ„¢ Flexible Fit Earplugs equipped with a probe tube for fit testing using the 3Mâ„¢ E-A-Rfitâ„¢ Dual-Ear Validation System. Cannot be used for hearing...
UOM: 1 * 50 PAIR
Supplier: 3M
Description: This foam earplug with tapered bell shape and super-soft foam is easy-to-fit and seals effectively to provide the highest NRR available in the industr...
Supplier: 3M
Description: These roll-down earplugs are made from a soft, energy-absorbing polymer foam which provides excellent hearing protection and all-day comfort.
Supplier: 3M
Description: These pod earplugs are very easy to insert since there is no roll needed, users can just use the soft flexible grip to push the earplug into position....
Supplier: 3M
Description: E-A-RSoftâ„¢ yellow neon earplugs are made from a slow expanding polyurethane foam material. A plug that gives evenly distributed pressure allows flexib...
Catalog Number: (PRTWEP08ORR)
Supplier: Portwest
Description: These disposable ear plugs are made from soft PU foam and PVC.
UOM: 1 * 200 PAIR
Catalog Number: (PRTWEP30BLU)
Supplier: Portwest
Description: These detectable disposable ear plugs are comfortable to fit due to the soft PU foam material. Corded for increased safety and visibility in food indu...
UOM: 1 * 200 items
Supplier: Moldex
Description: SparkPlugs® earplugs are made of a soft foam which becomes firmer when rolled for insertion.
Sale
Supplier: Moldex
Description: Earplugs made from soft PU-foam for protection against lower noise levels.
Supplier: Moldex
Description: These extremely comfortable earplugs made of soft polyurethane foam are available in two different sizes.
Catalog Number: (MOLD765002)
Supplier: Moldex
Description: The Moldex® MelLows® antimicrobial earplugs are supplied as a PlugStation with five-hundred pairs of ear plugs. Renowned for their hygienic hearing pr...
UOM: 1 * 500 PAIR
Supplier: Moldex
Description: SparkPlugs® earplugs are made of soft, extra light foam.
Supplier: Moldex
Description: These plugs made of soft PU foam provide protection against higher noise levels.
Safer Human and Environmental Health
Catalog Number: (MOLD776001)
Supplier: Moldex
Description: The classic Pura-Fit® earplugs offer the optimal combination of stability and softness. They are easy to roll and, upon insertion, expand to fill the ...
UOM: 1 * 500 PAIR
Supplier: UVEX
Description: Earplugs Com4-Fit, refill box for dispenser "one 2 click", uvex com4-fit, Disposable, SNR: 33 dB, Pink, In pairs
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 28
no targeter for Bottom