You Searched For: Earmuffs with Radio
10  results were found
SearchResultCount:"10"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: 3M
Description: 3Mâ„¢ PELTORâ„¢ WSâ„¢ LiteCom Pro III headsets are high-performance hearing protectors with a built-in analogue and digital programmable two-way communicati...
Supplier: 3M
Description: The Peltorâ„¢ WSâ„¢ LiteCom Plus Headset is available in multiple mounting styles, and features an integrated pre-programmed analog two-way radio, Bluetoo...
Supplier: 3M
Description: Peltor™ WS™ ProTac XPI is a range of level-dependent Bluetooth® headsets for effective hearing protection without compromising communication. They com...
Catalog Number: (MMMAMRX21A3WS6)
Supplier: 3M
Description: The Peltor™ WS™ Alert™ XPI Headset is a is high-attenuating hearing protector that can connect up to two Bluetooth® enabled devices, such as a mobile ...
UOM: 1 * 1 items
Supplier: 3M
Description: Peltorâ„¢ LiteCom Headsets are hearing protectors with a built-in, pre-programmed two-way communication radio and an integrated noise cancelling microph...
Catalog Number: (MMMAHRXS220A)
Supplier: 3M
Description: The headset helps to protect from potentially harmful workplace noise while at the same time the wearer can listen to the built in AM/FM radio.
UOM: 1 * 1 items
Catalog Number: (MMMAMT73H7P3E441EU)
Supplier: 3M
Description: Peltorâ„¢ LiteCom plus headsets are robust hearing protectors with a built-in analogue two-way communication radio, noise cancelling dynamic microphone,...
UOM: 1 * 1 items
Catalog Number: (MMMAM2RX7A)
Supplier: 3M
Description: This high-attenuating over-the-head hearing protection headset incorporates an electronic level dependent AM/FM radio receiver with approximately 70 h...
UOM: 1 * 1 items
Catalog Number: (111-2496)
Supplier: 3M
Description: The 3M™ Peltor™ WS™ LiteCom Pro III Headset features an integrated, programmable analog/digital two-way radio, Bluetooth® Multipoint connectivity, noi...
UOM: 1 * 1 items
Catalog Number: (DALL1030330)
Supplier: HONEYWELL SAFETY
Description: Comfortable and lightweight radio earmuffs featuring a superior digital AM/FM sound.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 10 of 10
no targeter for Bottom