Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?
  • Criteria
    • In Stock (16)
  • Supplier
  • Refine by Suppliers
    Sort by:
  • Search Within Results
Temperature Indicating Strips
https://ru.vwr.com/stibo/smallweb/std.lang.all/99/01/18989901.jpg
SearchPresentationType-VERTICAL
SearchResultCount:"10"
  List View Easy View (new)
Sort by:
Supplier:  VWR Collection
Description:   These temperature indicating strips are designed for rapid measurements where conventional thermometers are impractical. They are suitable for use wit...
Catalog Number: (BRDY195844)
Supplier:  Brady
Description:   These reversible temperature indicator labels designed to display temperatures of food and medical supply packaging or of components during research, ...
UOM:  1 * 10 items
Supplier:  TQC
Description:   These single temperature indicating stickers are designed for use in electronics, food and pharmaceutical industries, and also in the oven and engine ...
Supplier:  TQC
Description:   These temperature indicating strips are designed for use in electronics, food and pharmaceutical industries, and also in the oven and engine construct...
Supplier:  3M MEDICAL
Description:   Freeze watch indicators consist of a highly sensitive indicating liquid inside a specially designed ampoule to monitor exposure of temperature-sensiti...
Supplier:  3M MEDICAL
Description:   The MonitorMark™ temperature indicators incorporate both a temperature indication (10 °C) and a 34 °C GO/NO GO threshold indicator (34 °C). ...
Supplier:  Testo
Description:   These thermometer strips are made of self-adhesive foils with temperature sensitive elements for temperature control and regulation. They are used for...
Supplier:  Testo
Description:   These clock indicators are self-adhesive, temperature proof foils with temperature sensitive elements for temperature control and regulation. They are...
Call for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8  of 10
  1  2  Next 
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping