Pesticide reference standard, Trinexapac-ethyl, PESTANAL®, analytical standard, Supelco®
Supplier: Merck
PESTANAL®
37898-100MG-REA 82.2 EUR
37898-100MG-R
Pesticide reference standard, Trinexapac-ethyl, PESTANAL®, analytical standard, Supelco®
Standards Organic Standards
Trinexapac-ethyl, a compound that belongs to the cyclohexanedione group, is a growth regulator, which can find applications in agriculture and horticulture. Its mode of action basically involves the suppression of gibberellin biosynthesis.