Pesticide reference standard, Fenthion oxon, analytical standard, Supelco®
Supplier: Merck
89257-5MGEA 60.9 EUR
89257-5MG
Pesticide reference standard, Fenthion oxon, analytical standard, Supelco®
Standards Organic Standards
Used as a reference standard for the determination of fenthion oxon in animal tissue samples by gel permeation chromatography (GPC) cleanup followed by gas chromatography-mass spectrometry (GC–MS) and liquid chromatography coupled with tandem mass spectrometry (LC–MS–MS).