Pesticide reference standard, Carfentrazone-ethyl, PESTANAL®, analytical standard, Supelco®
Supplier: Merck
PESTANAL®
34079-10MGEA 151 EUR
34079-10MG
Pesticide reference standard, Carfentrazone-ethyl, PESTANAL®, analytical standard, Supelco®
Standards Organic Standards
Carfentrazone-ethyl is an aryl triazolinone reduced-risk herbicide used in the cultivation of wheat, soybean and corn. Carfentrazone-ethyl may be used as a reference standard in the determination of carfentrazone-ethyl in: Water samples using direct immersion solid-phase microextraction (DI-SPME) with gas chromatography coupled with mass spectrometry (GC-MS), Honey samples using liquid chromatography-electrospray ionization-tandem mass spectrometry (LC-ESI-MS/MS).