Ammonia (nitrogen) test strips
Supplier: Hach
HACH2755325EA 21.1 EUR
HACH2755325
Ammonia (nitrogen) test strips
Environmental Test Kits
Ammonia (Nitrogen) is a product of microbiological decay of plant and animal protein. Its presence in raw surface waters usually indicates domestic or agricultural pollution. Above certain levels, it is toxic to fish.
  • Easy to use, disposable, and inexpensive
  • A great way to obtain quick, quantitative answers in the field or in the lab
  • Parameter: ammonia, nitrogen, low range - As NH₃-N, for freshwater