Harness adaptor, DBI-SALA, 3M™
Supplier: 3M
CPSD1500161EA 11.9 EUR
CPSD1500161
Harness adaptor, DBI-SALA, 3M™
The DBI-SALA® harness adaptor is lightweight, versatile attachment point for a tool lanyards.
  • Fast and easy installation
  • Durable anodized aluminium construction
The harness adapter is designed to be used with DBI-SALA 2,2 kg tool retractor, Hook2Loop Bungee Tether, Hook2Hook coil tether with swivel, Hook2Hook coil tether and Hook2QuickRing coil tether with tail.
Capacity: 2,3 kg