You Searched For: Light Guides
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Light Guide, Light guide focus, VWR®, For: Fibre optic light guides, Focus optic for fibre optic light guide
Supplier: SCHOTT AG Lighting and imaging
Description: Light Guide, Flexible light guide, 1-branch, 1000 mm, Flexible light guide, 1-arm, Ø=4,5 mm/1000 mm
Supplier: SCHOTT AG Lighting and imaging
Description: Fibre optic illumination for stereo microscopy. To be used with cold light sources.
Supplier: SCHOTT AG Lighting and imaging
Description: Light Guide, Transmitted light stage TLS-BF Ø= 84 mm, Transmitted light stage TLS-BF Ø= 84 mm
Supplier: SCHOTT AG Lighting and imaging
Description: Light Guide, Lightline with adjustable front lens slit, 50×1,2 mm
Catalog Number: (LEIM31154202)
Supplier: LEICA MICROSYSTEMS
Description: High quality schott fiber optics for perfect light placement. Microsystemsâ„¢ Schott Gooseneck Light Guide provides flex and stay lighting.
UOM: 1 * 1 items
Supplier: MOTIC
Description: Light Guide, Gooseneck light guide, two arms, Ø×L: 0,5 mm×200 mm, Gooseneck light guide, two arms, Ø×L: 0,5 mm×200 mm
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom