Stacking containers, white
216-2205EA 43.5 EUR
VWRI216-2205
Stacking containers, white
Made of polypropylene, white.
  • Available in 5 different sizes
  • Can be nested for compact storage
  • Smooth interior walls make it easy to clean
  • Ergonomic handles for easy lifting
  • Use with temperatures of –25 to +80 °C
  • 100% recyclable
These stackable transport containers are ideal for use in storage, transport and food processing applications. Thanks to their conical shape, the containers can be nested into each other to save space. They have a smooth inner and are food safe.