Isobutanol, Supelco®
M-Clarity™ quality level = MQ100
82059-1ML-FEA 47.8 EUR
82059-1ML-F 82059-10ML-F 82059-5ML-F
Isobutanol, Supelco®
Isobutanol
Formula: (CH₃)₂CHCH₂OH
MW: 74,12 g/mol
Boiling Pt: 107,9 °C (1013 hPa)
Melting Pt: –108 °C
Density: 0,802 g/cm³ (20 °C)
Flash Pt: 28 °C
MDL Number: MFCD00004740
CAS Number: 78-83-1
UN: 1212
ADR: 3,III
Merck Index: 13,05148
https://export.vwr.com/stibo/web/std.lang.all/72/70/26837270.jpg