1-Heptanesulphonic acid sodium salt ≥98.0% (by acidimetry) for HPLC, for ion pair chromatography, J.T.Baker®
Supplier: Avantor
Warning
2173-05EA 203.06 EUR
2173-05
1-Heptanesulphonic acid sodium salt ≥98.0% (by acidimetry) for HPLC, for ion pair chromatography, J.T.Baker®
1-Heptanesulphonic acid sodium salt
For ion-pair chromatography of basic compounds. UV cutoff 210 nm.
Formula: H₃C(CH₂)₅CH₂SO₃Na
MW: 202,25 g/mol
Melting Pt: 300 °C
MDL Number: MFCD00007543
CAS Number: 22767-50-6
https://export.vwr.com/stibo/web/std.lang.all/07/91/2300791.jpg
Order Now
Specification Test Results
For Laboratory,Research,or Manufacturing Use
Assay (as C₇H₁₅SO₃Na) (by Acidimetry) ≥98.0%
UV Absorbance of 0.25 M Solution - 250 nm ≤0.050
UV Absorbance of 0.25 M Solution - 240 nm ≤0.050
UV Absorbance of 0.25 M Solution - 230 nm ≤0.050
UV Absorbance of 0.25 M Solution - 220 nm ≤0.060
UV Absorbance of 0.25 M Solution - 210 nm ≤0.080
UV Absorbance of 0.25 M Solution - 200 nm ≤0.200