Digital timer, vibrating, one channel, VWR®
609-0227EA 27.6 EUR
VWRI609-0227
Digital timer, vibrating, one channel, VWR®
This timer vibrates and flashes when time is up. It quietly nudges users during a meeting but is also perfect for loud environments when hearing a buzzer is impossible.
  • Timing capacity: 19 hours, 59 minutes, or 99 minutes, 59 seconds
  • Three alarm modes: Audible, flashing LED and vibrating
  • Countdown and alarms on a single channel; counts up in stopwatch mode
  • Automatic Bounceback™ memory for repetitive timing
  • Counts up during alarming indicating elapsed time since alarming
  • Settable to one second with a one-second resolution
Certifications: Complies with ISO/IEC 17025, accredited by A2LA. Indicates traceability to standards provided by NIST.
Delivery information: Supplied with stand, magnetic back, batteries and certificate.