VWR® kuroGEL Verti 1010 Dual-gel vertical electrophoresis systems
Supplier: VWR
VWR®
700-0167EA 633 EUR
700-0167
VWR® kuroGEL Verti 1010 Dual-gel vertical electrophoresis systems
Electrophoresis Systems Vertical Gel Electrophoresis Systems
The kuroGEL Verti vertical dual-gel units offer the flexibility of running dual gels either cooled or uncooled and with or without a casting base.
  • Novel clamping arrangement allows most commercially available 8×10 cm or 10×10 cm precast polyacrylamide gels in a wide range of cassette thicknesses to be run
  • Two screws make clamping simple without using separate clamps
  • Self-sealing snap-on connectors prevent the spills which commonly occur when disconnecting the cooling liquid
The kuroGEL Verti 1010 mini gel units are ideal for standard PAGE applications requiring self-cast gels. Soft silicone gaskets in the precision engineered casting base act in tandem with the gel running module to provide simple, leak-free casting and assembly.
Delivery information: Complete system supplied with 2 plain glass plates; 2 notched glass plates; 1 dummy plate; 2×1,0 mm spacer sets; 2×1,0 mm combs, 12-sample wells.