Silicone lubricant and sealant, Molykote™ 111
Supplier: Dow
636052SEA 53.3 EUR
636052S
Silicone lubricant and sealant, Molykote™ 111
Lubricants
Molykote 111 is a valve lubricant and sealant with good resistance to most chemicals and a wide service temperature range.
  • Wide service temperature range (40 to 200 °C)
  • Low vapour pressure
  • Low volatility
  • Excellent water resistance