Microarray Slides, GAPS™ II (Corning®)
Supplier: Corning
734-1684EA 525.65 EUR
CORN40006 CORN40004 CORN40005 CORN40003
Microarray Slides, GAPS™ II (Corning®)
  • Recommended for both DNA and protein arrays
  • High DNA retention for maximum signal strength
  • Low background autoflourescence