Radio-digital thermo-hygrometer, Traceable®
620-1674EA 95.2 EUR
VWRI620-1674
Radio-digital thermo-hygrometer, Traceable®
This wireless thermo-hygrometer, readable from up to 30 metres (433 MHz), can receive and display data from up to three remote modules. They can be connected to one base unit, which displays the temperature and humidity of the base unit and of the three different remote modules.
  • Data transmitted every 30 seconds
  • Trend indicators: Rising or falling
  • Maximum/minimum display
  • Audible alarms on modules
  • Jumbo digits
  • IP 20
Certifications: Traceability to standards provided by NIST. Compliance with ISO/IEC 17025, ANSI/NCSL Z540-1 and 9001.
Delivery information: Supplied with remote sensor module and Traceable® certificate of calibration.
RH = Relative Humidity