LC-MS nitrogen generators, Parker Domnick Hunter
Supplier: Parker
Parker domnick hunter
564-0016EA 11516 EUR
564-0016 564-0015 564-0106 564-0017 564-0105 564-0093
LC-MS nitrogen generators, Parker Domnick Hunter
Gas Generators Nitrogen Generators
Parker Domnick Hunter LC-MS nitrogen generators are specifically designed to meet the gas flow, purity and pressure requirements of the latest generation of LC-MS instruments. Using the pressure swing adsorption technique, these generators provide a continuous stream of clean, dry nitrogen without the need for secondary purification, and are fully compatible with all known APCI and ESI interfaces.
  • Suitable for nebulising, sheath and drying gas
  • Fully regenerative, proven durability PSA technology
  • Phthalate-free components
Models featuring integral oil-free compressors give increased security of supply, completely removing the reliance on external air supplies.
To ensure optimum performance and longevity, we recommend the use of maintenance kits.
Contact Avantor for your installation and service requirements.