Freezing module, CoolCell®
479-0492EA 170 EUR
BCISBCS-136
Freezing module, CoolCell®
Consistent -1 °C/min cell freezing without alcohol
  • No alcohol
  • Eliminate the expense and disposal of isopropanol
  • Unbreakable even at cryogenic temperatures
  • No stuck lids, no frozen fingers, no maintenance
  • Fast recycle time
Achieve a consistent -1 °C/min freeze rate without the use of any hazardous solvents with CoolCell®. The Solid State Core (SSC) and insulated design precisely balance heat removal during freezing to ensure repeatable, constant cooling all the way down to cryogenic storage temperature. Cleaner, greener, easier to use. Unlike alcohol coolers, no warming of nearby freezer samples. Transportable without cold hands when frozen.