You Searched For: Faucets and Sinks
20  results were found
SearchResultCount:"20"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (29048.098)
Supplier: VWR Chemicals
Description: PE, vented, for use with plastic jerricans with external thread (DIN 42).
UOM: 1 * 1 items
Catalog Number: (332862K)
Supplier: VWR Chemicals
Description: PE, white, ³/₄" screw thread.
UOM: 1 * 1 items
Catalog Number: (331582B)
Supplier: VWR Chemicals
Description: PP, white, ³/₄" screw thread.
UOM: 1 * 1 items
Supplier: KOETTERMANN
Description: These bench mounted gas taps are sealed by EPDM (ethylene propylene diene terpolymer) which is an extremely durable synthetic rubber roofing membrane.
Supplier: asecos
Description: These drum taps are made of stainless steel.
Supplier: KOETTERMANN
Description: Swivel water taps are sealed by EPDM (ethylene propylene diene terpolymer) or ceramic.
Catalog Number: (MSGA2796)
Supplier: asecos
Description: This fine dosing tap is made of zinc injection moulding.
UOM: 1 * 1 items
Supplier: S.C.A.T.
Description: For integration in fume cupboards or on worktops for easy disposal of liquid solvent waste in large quantities.
New Product
Catalog Number: (BOHLR1250-20)
Supplier: Bohlender
Description: Made of static dissipative PE-EX, with sloped bottom for complete emptying.
UOM: 1 * 1 items
Supplier: Lenz Laborglas GmbH & CO.KG
Description: These drain taps are made of borosilicate glass and are used for aspirator bottles.
Catalog Number: (BOHLR1250-40)
Supplier: Bohlender
Description: Made of static dissipative PE-EX, with sloped bottom for complete emptying.
UOM: 1 * 1 items
Supplier: BROEN-LAB
Description: UniFlexâ„¢ BASIC fittings for special water are designed to secure maximum purity of the water in laboratory. The operation and configuration of the fit...
Supplier: JUSTRITE MANUFACTURING CO
Description: These faucets can be simply placed on your drum for leakproof dispensing of flammable liquids. The brass construction with a polytetrafluoroethylene (...
Catalog Number: (BROE25350101035)
Supplier: BROEN-LAB
Description: BROEN®, One short handle two hole mixer with swivel spout, 200 mm
UOM: 1 * 1 items
Supplier: TEKNOMEK
Description: Our range of sensor operated wash troughs are manufactured from 304 grade stainless steel with a fully welded design for maximum hygiene. Choose from ...
Supplier: TEKNOMEK
Description: Stainless steel sink with double cupboard and drawer is manufactured from high quality, 304 grade stainless steel and is supplied with an inset sink a...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 20
no targeter for Bottom