You Searched For: Microtomes and Microtome Assemblies
8  results were found
SearchResultCount:"8"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: LEICA HISTOLOGY
Description: With these blades it is easy to section a wide range of tissues. Edge encourages immediate ribboning. Premium edge geometry with 35° angle, so no adju...
Catalog Number: (720-1964)
Supplier: Slee
Description: The CUT 4062 is a manual rotary microtome equipped with maintenance-free caged roller bearing guide ways and micrometre feed mechanism to provide the ...
UOM: 1 * 1 items
Catalog Number: (631-0561)
Supplier: Slee
Description: The MEV floorstanding cryostat features convenient operation and a modern rotary microtome with motorised μm advance/return. The space-saving design i...
UOM: 1 * 1 items
Supplier: Epredia
Description: Choose from HP35, MB22, or MX35 microtome blades.
Catalog Number: (631-0542)
Supplier: Slee
Description: The CUT 5062 is a semi-automated rotary microtome designed for paraffin sections.
UOM: 1 * 1 items
Catalog Number: (631-0545)
Supplier: Slee
Description: The CUT 6062 is a fully automated rotary microtome equipped with maintenance-free caged roller bearing guide ways and micrometre feed mechanism to pro...
UOM: 1 * 1 items
Supplier: Slee
Description: This instrument is equipped with a fully automatic rotary microtome with extremely high mechanical stability, tolerance-free roller guides, and state ...
Catalog Number: (PERS72-0114-0000)
Supplier: PERSONNA MEDICAL
Description: Personna Plus® microtome blades with MicroCoat® coating.
UOM: 1 * 50 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 8 of 8
no targeter for Bottom