You Searched For: Stylus and Scribers
3  results were found
SearchResultCount:"3"
Sort Results
List View Easy View (new)
Rate These Search Results
Catalog Number: (SIMPM795-1)
Supplier: Simport Scientific
Description: The microscope diamond slide writer is compatible with a wide variety of slide labellers.
UOM: 1 * 6 items
Catalog Number: (811-0103)
Supplier: Bel-Art Products, a Part of SP
Description: The Glascribe® Pen, with retractable tungsten carbide tip, lets you inscribe on glass, ceramics, and plastic.
UOM: 1 * 1 items
Catalog Number: (BELAF441500005)
Supplier: Bel-Art Products, a Part of SP
Description: Battery powered engraving tool to permanently mark most materials including glass, plastic, wood, metal, bone, and stone.
UOM: 1 * 1 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 3 of 3
no targeter for Bottom