Group lock boxes, portable, extra large
Supplier: Brady
BRDY830929EA 90.03 EUR
BRDY830929
Group lock boxes, portable, extra large
This extra-large metal lock box is made of heavy-duty steel and is powder coated for extra rust resistance. It can be used to assure all employees who are servicing the equipment that the energy isolation points will remain locked out until they've completed their work and removed their personal lockout devices.
  • Made of mild sheet steel
  • Accommodates up to 12 padlocks (locks not included)
  • Can also be used as a lock storage box holding up to 75 padlocks
  • Slot in front allows keys to be inserted when box is locked