You Searched For: Gaskets
29  results were found
SearchResultCount:"29"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: Avantor Fluid Handling
Description: Easily detect lost elastomeric fragments to prevent contamination and product recalls.
Supplier: Avantor Fluid Handling
Description: Hands-free installation for difficult-to-reach locations!
Supplier: Avantor Fluid Handling
Description: Ensure a tight seal.
Supplier: Avantor Fluid Handling
Description: Ensure a better process seal with outstanding chemical resistance.
Supplier: Avantor Fluid Handling
Description: Protect your pump, valves and other components in fluid line.
Supplier: Avantor Fluid Handling
Description: Create a secure connection with your Masterflex® Smart Gasket® sanitary gaskets.
Sale
Catalog Number: (MFLX30542-20)
Supplier: Avantor Fluid Handling
Description: Easily sample fluid in critical high-purity system.
UOM: 1 * 1 items
Supplier: Avantor Fluid Handling
Description: Smart Gasket lets you take samples or readings from a sanitary line.
Supplier: Avantor Fluid Handling
Description: Protect your pump, valves and other components in fluid line.
Supplier: Avantor Fluid Handling
Description: Protect your pump, valves and other components in fluid line.
Supplier: Avantor Fluid Handling
Description: Protect your pump, valves and other components in fluid line.
Supplier: Avantor Fluid Handling
Description: Greater particulate capacity for higher particulate flow.
Supplier: Avantor Fluid Handling
Description: Protect your pump, valves and other components in fluid line.
Supplier: Avantor Fluid Handling
Description: Simple accessories to protect your delicate pressure gauge.
Catalog Number: (MFLX30544-20)
Supplier: Avantor Fluid Handling
Description: Easily sample fluid in critical high-purity system.
UOM: 1 * 1 items
Supplier: Avantor Fluid Handling
Description: The benefits of PTFE and 316L stainless steel in one gasket!
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 29
no targeter for Bottom