Human plasma, colorectal cancer (CRC)
Supplier: Charles River Laboratories Cell Solutions
HCMSPB006C-1-CRCEA 0 EUR
HCMSPB006C-1-CRC
Human plasma, colorectal cancer (CRC)
Human peripheral blood plasma used for researching the causes, diagnosis, supportive care and treatment of colorectal cancer (CRC). Collected from IRB-consented donors diagnosed with colorectal cancer.
No pre-screening is done on fresh products, but donors are tested on day of collection for Hepatitis B, Hepatitis C, and HIV 1/2.
Storage: Store at −80 °C.
Caution: Intended for only research use and not for direct therapeutic or diagnostic use in human or animals, or for further manufacturing of therapeutic products or devices.