Seed Direct PCR Kits
Supplier: Omega Bio-Tek
TQ2320-00EA 40.8 EUR
OMEGTQ2320-00 OMEGTQ2320-01
Seed Direct PCR Kits
Seed Direct PCR Kit offers a quick and simple procedure to extract genomic DNA from seeds for PCR.
For best possible yield of DNA from seed samples, a longer incubation time at 55 °C may be necessary. This is due to variations such as seed size and moisture content.