Filter funnel/column, with standard ground joint and filter disc, SP Bel-Art
Supplier: SP Industries
WILMLG-4537-138EA 363 EUR
WILMLG-4537-138
Filter funnel/column, with standard ground joint and filter disc, SP Bel-Art
Made of borosilicate glass.
  • Very good temperature resistance
  • Very good chemical resistance
  • Funnel has an standard taper inner joint at the bottom with a coarse porosity frit
These funnel/columns are used where rapid separation is needed.
Height (above disc): 101,6 mm