Upright microscope, Panthera L
Supplier: Motic
Panthera
MOTI1100104600142EA 0 EUR
MOTI1100104600142
Upright microscope, Panthera L
Panthera L is a Life science model standing out for its combination of build-in digital capabilities.
  • HDMI, Wi-Fi, LAN and USB for external devices
  • Plan achromatic UC objectives (ultra-high contrast) for bright and sharp images
  • Sleep mode for lab safety and saving energy
  • Motic Light Tracer includes coded nosepiece, LED indicator of light intensity, and digital knob
  • Contrasting techniques include: Brightfield, darkfield, phase contrast, polarisation and EpiLED fluorescence
Its SMART CAM digital head and the imaging on-device system including a combination of software and hardware allows the user to work from microscope to an HDMI screen with a mouse, or work with tablet and the Panthera App.
Delivery information: Includes a blue filter, immersion oil (5 ml), power cable, dust cover, Allen key, halogen bulb and LED module.