Sound level meter, Testo 816
Supplier: Testo
TEST0563.8165EA 390 EUR
TEST0563.8165
Sound level meter, Testo 816
The testo 816 meter with AC/DC output for data readout is the professional model in the Testo range of sond measuring instruments. It is designed specifically for measuring sound levels in the workplace, assessors and for measuring industrial and environmental noise.
  • Bar graph display
  • Automatic range switchover
  • Battery life: 50 hr
  • Easy to adjust
  • Switchable time weighting fast/slow
  • DC output 10 mV/dB for connecting recorders
  • Frequency evaluation according to characteristic curves A and C
Via the AC output, recorders or amplifiers can be connected directly to the instrument, and via the DC output, graphical recorders or data loggers can be used easily and directly on site.
Delivery information: Includes microphone, wind protection cap, battery, stereo jack 3,5 mm in system case with battery.