You Searched For: Ceramic Beads
7  results were found
SearchResultCount:"7"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Grinding ball, Set of 5 pieces, Zirconium oxide
Catalog Number: (33190.A1)
Supplier: Thermo Scientific
Description: Grinding pieces, Al-23, 10 mm, 440 pieces/kg, Ceramic
UOM: 1 * 1 kg
MSDS
Supplier: Thermo Scientific
Description: Pure Al₂O₃ Aluminium oxide, AL 23.
Supplier: Thermo Scientific
Description: The density of the zirconium oxide beads is 6.02 g/cm³
Supplier: MP Biomedicals
Description: Ceramic sphere is a Zirconium oxide coated grinding matrix which is chemically inert and will not bind nucleic acids. These are components of lysing m...
Supplier: LEMKE, GERD
Description: Agate.
Supplier: SIGMUND LINDNER GMBH
Description: Ceramic beads made of Zirconium Oxide / Yttrium stabilised.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 7 of 7
no targeter for Bottom