You Searched For: Ultra Low Temperature Freezers
17  results were found
SearchResultCount:"17"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: VWR® ULT Eco Premium freezers are designed for safe and reliable storage of sensitive samples. They feature ergonomic control and low energy consumpti...
Energy Efficient
Low Manufacturing Impact Sale
Supplier: Thermo Scientific
Description: Thermo Scientificâ„¢ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models with 5 Inner doors, maximizing storage c...
Sustainable Materials
Supplier: Thermo Scientific
Description: Thermo Scientific™ TDE Series ‒40 °C ultra-low temperature freezers, powered by H-drive, feature three upright models, maximizing storage capacity fro...
Sustainable Materials
Supplier: REVCO TECHNOLOGIES
Description: Thermo Scientificâ„¢ Revcoâ„¢ RDE Series ultra-low freezers feature four upright models, maximizing storage capacity from 30,000 up to 60,000 2 ml vials. ...
Energy Efficient
Safer Human and Environmental Health
Catalog Number: (REVCRDE50040FV)
Supplier: REVCO TECHNOLOGIES
Description: The RDE series of ultra low temperature freezers have been designed to keep your samples safe. Worry-free storage; with low peak variation, samples a...
UOM: 1 * 1 items
Supplier: Thermo Scientific
Description: Thermo Scientificâ„¢ TDE Series ultra-low temperature chest freezers set the standard for performance and reliable sample storage in heavy use situation...
Sustainable Materials
Supplier: Thermo Scientific
Description: Thermo Scientificâ„¢ TSX series ultra-low high performance freezers are designed with features that support sustainability objectives without compromisi...
Supplier: Thermo Scientific
Description: TSX Universal Series ULT Freezers are the pinnacle of reliability, sustainability, and innovation incorporating insights from over 80 years of experie...
New Product
Supplier: REVCO TECHNOLOGIES
Description: Thermo Scientificâ„¢ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models with five inner doors, maximising storag...
Sustainable Materials
Supplier: Thermo Scientific
Description: Thermo Scientific™ TDE Series ‒40 °C ultra-low temperature freezers, powered by H-drive, feature three upright models with 5 inner doors, maximizing s...
Sustainable Materials
Catalog Number: (471-1238)
Supplier: Stirling Ultracold
Description: Ultra-low temperature freezer featuring the free-piston Stirling engine. This next generation of environmentally sustainable ULT freezers achieve stab...
UOM: 1 * 1 items
Supplier: Thermo Scientific
Description: Thermo Scientificâ„¢ TDE Series ultra-low temperature freezers, powered by H-drive, feature four upright models, maximizing storage capacity from 30000 ...
Sustainable Materials
Supplier: REVCO TECHNOLOGIES
Description: Thermo Scientificâ„¢ TDE Series ultra-low temperature chest freezers set the standard for performance and reliable sample storage in heavy use situation...
Sustainable Materials
Catalog Number: (471-1240)
Supplier: Stirling Ultracold
Description: The ULT25NEU is the only portable ULT freezer solution available today for bringing ULT storage to the patient. This small lightweight, and efficient...
UOM: 1 * 1 items
Catalog Number: (471-1239)
Supplier: Stirling Ultracold
Description: The SU105UE is a compact ultra-low temperature freezer that is ideal for personal storage and situations where larger volume ULT freezers might otherw...
UOM: 1 * 1 items
Supplier: Binder
Description: These ultra-low temperature freezers ensure the safe storage of samples at –90 °C. They combine outstanding environmental friendliness with low energy...
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 17
no targeter for Bottom