You Searched For: Scalpel Handles
18  results were found
SearchResultCount:"18"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: SWANN-MORTON
Description: Stainless steel.
Catalog Number: (SWAN5398)
Supplier: SWANN-MORTON
Description: This stainless steel handle is designed to assist the OR nurse with safe and easy blade attachment and removal before, during and after the procedure....
UOM: 1 * 1 items
Supplier: SWANN-MORTON
Description: These handles and blades are ideal for artwork, craft work and modelling.
Supplier: SWANN-MORTON
Description: Stainless steel.
Supplier: SWANN-MORTON
Description: The heavy duty handles are made from stainless steel.
Supplier: SWANN-MORTON
Description: These acrylic craft knife handles have a stainless blade fitting.
Supplier: SWANN-MORTON
Description: A versatile and comprehensive range for morticians and pathologists for use in autopsy, histopathology and general post-mortem procedures.
Catalog Number: (233-5461)
Supplier: SWANN-MORTON
Description: Stainless steel, non sterile.
UOM: 1 * 1 items
Catalog Number: (SWAN6061)
Supplier: SWANN-MORTON
Description: Stainless steel.
UOM: 1 * 1 items
Supplier: SWANN-MORTON
Description: Bulbous stainless steel handles for surgical scalpel blades. Individually sleeved.
Catalog Number: (SWAN1235)
Supplier: SWANN-MORTON
Description: Handle made from impact resistant PS.
UOM: 1 * 10 items
Supplier: KLS Martin Group
Description: These scalpel handles are made from stainless steel.
Supplier: HAMMACHER KARL
Description: Stainless steel.
Catalog Number: (233-5020)
Supplier: HAMMACHER KARL
Description: Solid scalpel handle made from stainless steel. For use with blades HSB 913-20 to HSB 935-12.
UOM: 1 * 1 items
Supplier: BAYHA GMBH
Description: Stainless steel.
Supplier: AESCULAP AG & CO
Description: Scalpel handle, Aesculap®, Carbon steel, No. 4, For: Blades 18 - 36, Length: 135 mm, Scalpel handle, short, carbon steel
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 18
no targeter for Bottom