You Searched For: Scalpel Blades
24  results were found
SearchResultCount:"24"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: SWANN-MORTON
Description: Scalpel blades, Carbon steel, Sterile, No. 15T, Individual foil packs, carbon steel
Supplier: SWANN-MORTON
Description: Stainless steel.
Supplier: SWANN-MORTON
Description: Blades made from carbon steel for use with craft tools.
Supplier: SWANN-MORTON
Description: These stainless steel blades are thicker than a standard blade thus offering additional rigidity and strength when used in conjunction with the unique...
Supplier: SWANN-MORTON
Description: Stainless steel.
Catalog Number: (SWAN0326)
Supplier: SWANN-MORTON
Description: The stainless steel No.3 fitment or 'baby' stitch cutter is used in conjunction with the No.3 surgical handle.
UOM: 1 * 100 items
Supplier: SWANN-MORTON
Description: Carbon steel.
Supplier: SWANN-MORTON
Description: These Dermatome blades are fitted into a handheld instrument which resembles an electric razor. The blade oscillates and the movement back and forth w...
Supplier: SWANN-MORTON
Description: Surgical quality blades for use with handles No. 5A and No. 6A.
Supplier: SWANN-MORTON
Description: These Sabre blades made from carbon steel feature a 'closed' cutting edge for 'paring' or 'scraping' which makes them ideal for use within Podiatry an...
Supplier: SWANN-MORTON
Description: Stainless steel. The 158 mm skin graft blades are used in the surgical removal of a patch of healthy skin from one area of the patient's body, usually...
Catalog Number: (SWAN9942)
Supplier: SWANN-MORTON
Description: This custom designed surgical stainless steel Silver's blade has a 11/2 inch (4 cm) postage stamp size cutting edge to be used with the Silvers miniat...
UOM: 1 * 10 items
Catalog Number: (233-0053)
Supplier: SWANN-MORTON
Description: Stainless steel. Originally developed for use with skin grafts, these blades are also suitable for a wide range of other applications.
UOM: 1 * 20 items
Supplier: SWANN-MORTON
Description: A versatile and comprehensive range for morticians and pathologists for use in autopsy, histopathology and general post-mortem procedures.
Catalog Number: (SWAN2102)
Supplier: SWANN-MORTON
Description: The sterile stainless steel Myringotomy blade is used in conjunction with the SF1 or SF13 Fine surgical handle in the treatment of 'Otitis media' or '...
UOM: 1 * 10 items
Catalog Number: (SWAN2001)
Supplier: SWANN-MORTON
Description: This sterile stainless cervical biposy blade is used in conjunction with the long stainless steel Major handle to take a 'cone' biopsy from the tissue...
UOM: 1 * 10 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 24
no targeter for Bottom