Bottle top dispensers, Zippette™ Classic
Supplier: VWR Collection
613-0926EA 294 EUR
JENC263-000 JENC263-001 JENC263-002 JENC263-003 JENC263-004
Bottle top dispensers, Zippette™ Classic
Variable volume, autoclavable.
  • Fine adjustment for exact and reproducible dispensing
  • All wetted parts are chemically resistant and autoclavable at 121 °C
  • Easily removable PTFE piston for smooth action and easy cleaning
  • Borosilicate glass barrel protected with a transparent PP sleeve can be easily removed and cleaned
  • Easy priming and minimum waste with no leakage back into the reservoir - the Zippette™ stays fully primed all day
Accurate dispensing from an easy to use dispenser. The specially designed delivery spout ensures exact dispensing into narrow neck containers and the anti-drip safety valve makes sure the Zippette™ does not leak when not in use.
Delivery information: The Zippette™ Classic dispensers have a standard 30 mm neck fitting and are supplied with three bottle neck adapters (38, 40 and 45 mm), and an individual calibration certificate. Please note that the Zippette™ Classic are not supplied with a reservoir.