You Searched For: Adhesive Slides
25  results were found
SearchResultCount:"25"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: Silane coated (aminoalkylsilane) glass slides producing a sticky surface, which can bind tissue sections to glass.
Supplier: VWR Collection
Description: Microscope slides recommended for haematoxylin and eosin (H&E) stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopatholog...
Sale
Catalog Number: (KLINKP-SIL-3059)
Supplier: KLINIPATH
Description: Microscope slides for slide printers.
UOM: 1 * 2.500 items
Supplier: VWR Collection
Description: Superfrost Plus slides are specially treated for frozen tissue sections.
Catalog Number: (631-1162)
Supplier: VWR Collection
Description: Silane (aminoalkylsilane) coating provide permanent adhesion, which can bind tissue sections to glass slides with a sticky surface.
UOM: 1 * 2.500 items
Catalog Number: (631-1238)
Supplier: KLINIPATH
Description: These silane adhesive slides are specially designed for thermal transfer printers.
UOM: 1 * 2.500 items
Catalog Number: (631-0107)
Supplier: VWR Collection
Description: These slides are coated with a permanent adhesive that provides excellent cell and tissue adhesion.
UOM: 1 * 72 items
Catalog Number: (631-1237)
Supplier: KLINIPATH
Description: This microscope slide is designed with grounded-90, finished and clipped corners of 1 mm.
UOM: 1 * 72 items
Supplier: Epredia
Description: Superfrost® Plus slides hold tissue sections without the need for expensive coating steps.
Supplier: LABONORD
Description: These slides can be used for the diagnostic testing of cells suspension and tissue samples.
Sale
Catalog Number: (630-2184)
Supplier: LEICA HISTOLOGY
Description: These adhesive slides are coated for a positive charge to enhance hydrophilicity.
UOM: 1 * 72 items
Catalog Number: (631-1349)
Supplier: Epredia
Description: These slides, made using a technology that electrostatically and chemically attracts fresh cell preparations, formalin-fixed Bouin's or alcohol-fixed ...
UOM: 1 * 72 items
Supplier: MARIENFELD
Description: The glass surface of the HistoBond® slide is loaded with a permanent positive charge, which fastens all types of section to the glass.
Supplier: MARIENFELD
Description: Adhesion slides with frosted ends and 90° ground edges.
Catalog Number: (630-2092)
Supplier: LABONORD
Description: These slides can be used for the diagnosis of suspensions of cells and samples of tissue and are suitable for in vitro diagnostics.
UOM: 1 * 72 items
Catalog Number: (MARI0810701)
Supplier: MARIENFELD
Description: HistoBond slides combine frosted ends with printed in various colours.
UOM: 1 * 2.000 items
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 25
no targeter for Bottom