Powder containment cabinets, XIT
Supplier: Bigneat
XIT
135-1682EA 2840 EUR
135-1682 135-2024 135-1683 135-0657
Powder containment cabinets, XIT
Fume Hoods and Enclosures Extraction Cabinets
For protection from particles whilst weighing and handling powders. Two-stage filtration is used in these particulate filtration systems. Firstly a pre-filter removes particles >5 µm, then a high capacity design HEPA filter, certified to BS EN1822, removes particulate matter >0,3 µm to an absolute filtration of 99,997%.
F3 XIT: Small weighing cabinets:
The mild steel fan housing is epoxy coated
Transparent panels (6 mm) are manufactured from clear acrylic which is resistant to corrosion and easy to clean
Ideal for a single balance, this cabinet has very small footprint
Compact size and portable
Fan speed provides for optimum face velocities of between 0,3 to 0,5 msˉ¹
F3 XIT cabinets feature hinged access flap
XIT Plus cabinets: Weighing cabinets:
The mild steel fan housing is epoxy coated
Transparent panels are fabricated from clear acrylic which is resistant to corrosion and easy to clean
Keypad control panel with touch button controls and service due indicator
Audible electronic low air flow alarm
Safechange pre-filter bagged under negative pressure
Variable fan-speed control provides for optimum face velocities; 0,3 to 0,5 msˉ¹
Vaneometer indicates flow condition.
Ordering information: Base trays are supplied with XIT Plus models. It is optional for F3 XIT-D, if required, it must be ordered separately – please contact us for more details.
Visit by qualified engineer to deliver, assemble, commission and air flow test powder weighing, ductless fume or PCR cabinets. Advice to user(s) of best practice.
Ordering information: Base trays are supplied with XIT Plus models. It is optional for F3 XIT-D, if required, it must be ordered separately – please contact us for more details.