This vacuum system is especially suitable for controlling processes in a rotary evaporator. The patented, closed system arrangement enables gentle distillation and high recovery rates of solvents even with low-boiling points. Ideal for use with the VWR® RV 8 and RV 10 rotary evaporators.
- Gentle distillation due to a closed system
- Reliable control of the evaporation process
- High degree of solvent recovery
- The vacuum pump is gas-tight, 100% oil-free, maintenance-free and quiet
- Environmentally friendly
Delivery information: Includes the chemically resistant vacuum pump, base plate, separator, condenser, fine tuning valve, digital vacuum gauge, safety and venting valve, mains switch and mains cable.
Caution: * Flow rate at atmospheric pressure