VWR® VP 8 vac Chemically Resistant Vacuum Systems for Distillation
Supplier: VWR
VWR®
181-0156EA 0 EUR
181-0156 181-0157 181-0158
VWR® VP 8 vac Chemically Resistant Vacuum Systems for Distillation
Pumps Vacuum Diaphragm Pumps
This vacuum system is especially suitable for controlling processes in a rotary evaporator. The patented, closed system arrangement enables gentle distillation and high recovery rates of solvents even with low-boiling points. Ideal for use with the VWR® RV 8 and RV 10 rotary evaporators.
  • Gentle distillation due to a closed system
  • Reliable control of the evaporation process
  • High degree of solvent recovery
  • The vacuum pump is gas-tight, 100% oil-free, maintenance-free and quiet
  • Environmentally friendly
Delivery information: Includes the chemically resistant vacuum pump, base plate, separator, condenser, fine tuning valve, digital vacuum gauge, safety and venting valve, mains switch and mains cable.
Caution: * Flow rate at atmospheric pressure