You Searched For: Spill Control Materials
203  results were found
SearchResultCount:"203"
Sort Results
List View Easy View (new)
Rate These Search Results
Supplier: VWR Collection
Description: VWR® absorbents and spill-containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accide...
Sale
Catalog Number: (121-0194)
Supplier: VWR Collection
Description: This portable, economy spill kit comes in a high visibility, water-resistant, yellow PVC bag with handles. Designed for small spills of chemicals and ...
UOM: 1 * 1 KIT
Supplier: VWR Collection
Description: VWR® absorbents and spill containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accide...
Sale
Supplier: VWR Collection
Description: VWR® absorbents and spill containment products help keep your employees and work environment safe from unexpected spills, leaks, drips or other accide...
Sale
Supplier: VWR Chemicals
Description: Developed to economically treat small spills, such as those that can occur in a laboratory environment. Provides easy access to the appropriate contro...
Supplier: VWR Collection
Description: These economy spill kits combine absorbents and instructions in a space-saving container. They are ideal for workers on the go like truck drivers, fie...
Sale
Catalog Number: (332377C)
Supplier: VWR Chemicals
Description: These coarse granules are easy to use, and when applied to liquid spillages, will absorb up to 80% of their own weight.
UOM: 1 * 2,5 kg
Supplier: VWR Chemicals
Description: These refill sets can be used with five types of SPILL-X® treatment kits. They are available in different formats.
Catalog Number: (121-0196)
Supplier: VWR Collection
Description: This speciality spill kit contains sorbents, neutralising agent, PPE and accessories so that you are prepared in an emergency. Contained in a handy tu...
UOM: 1 * 1 KIT
Catalog Number: (121-1050)
Supplier: VWR Collection
Description: Spill kit suitable for dangerous liquids.
UOM: 1 * 1 KIT
Catalog Number: (129-0199)
Supplier: VWR Chemicals
Description: Set for absorption of organics, aqueous solutions, alkalis and acids. It contains.
UOM: 1 * 1 SET
Supplier: CATALYST KITTING SOLUTIONS
Description: Protective clothing, tools, absorbents and wipes arranged in four multifunctional kits, for rapid and safe cleaning of cytotoxic drug spillages. For u...
Catalog Number: (121-0197)
Supplier: VWR Collection
Description: This speciality spill kit contains sorbents, neutralising agent, PPE and accessories so that you are prepared in an emergency. Contained in a handy tu...
UOM: 1 * 1 KIT
Catalog Number: (99910.414)
Supplier: VWR Chemicals
Description: Algosol is a unique, natural product made from seaweed, suitable for the neutralisation of acids, such as hydrochloric, sulphuric, nitric and hydroflu...
UOM: 1 * 10 kg
Catalog Number: (121-0195)
Supplier: VWR Collection
Description: This speciality spill kit contains sorbents, neutralising agent, PPE and accessories so that you are prepared in an emergency. Contained in a handy tu...
UOM: 1 * 1 KIT
Supplier: Brady
Description: These extra-tough oil only pads absorb oil and petroleum based liquids.
Inquire for Price
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at +43 1 97002 - 0.
Dual use goods can only be delivered within the European Union.
Dual use goods can only be delivered within the European Union.
This product has been blocked by your organization. Please contact your purchasing department for more information.
The original product is no longer available. The replacement shown is available.
This product is no longer available. Alternatives may be available by searching with the VWR Catalog Number listed above. If you need further assistance, please call VWR Customer Service at +43 1 97002 - 0.
1 - 16 of 203
no targeter for Bottom